missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18027-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the transient receptor potential protein family, which is a diverse group of proteins with structural features typical of ion channels. This protein plays an important role in taste transduction, and has characteristics of a calcium-activated, non-selective cation channel that carries Na+, K+, and Cs+ ions equally well, but not Caions. It is activated by lower concentrations of intracellular Ca, and inhibited by higher concentrations. It is also a highly temperature-sensitive, heat activated channel showing a steep increase of inward currents at temperatures between 15 and 35 degrees Celsius. This gene is located within the Beckwith-Wiedemann syndrome critical region-1 on chromosome 11p15.5, and has been shown to be imprinted, with exclusive expression from the paternal allele.Specifications
| TRPM5 | |
| Polyclonal | |
| Unconjugated | |
| TRPM5 | |
| LTrpC 5, LTRPC5, MTR1, TRPM5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29850, 365391, 56843 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JJH7, Q9NZQ8 | |
| TRPM5 | |
| TRPM5 Fusion Protein Ag12593 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction