Learn More
Invitrogen™ TRPM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588498
Description
Immunogen sequence: RVFKKEAEHK REHLERDLPD PLDQKVVTWE TVQKENFLSK MEKRRRDSEG EVLRKTAHRV DFIAKYLGGL REQEKRIKCL ESQINYCSVL VSSVADVLAQ GGGPRSSQHC GEGSQLVAAD HRGGLDGWEQ PGAGQPPSDT; Positive Samples: U-87MG, Mouse liver, Mouse brain; Cellular Location: Cell membrane, Multi-pass membrane protein.
This gene encodes a member of the transient receptor potential protein family, which is a diverse group of proteins with structural features typical of ion channels. This protein plays an important role in taste transduction, and has characteristics of a calcium-activated, non-selective cation channel that carries Na+, K+, and Cs+ ions equally well, but not Caions. It is activated by lower concentrations of intracellular Ca, and inhibited by higher concentrations. It is also a highly temperature-sensitive, heat activated channel showing a steep increase of inward currents at temperatures between 15 and 35 degrees Celsius. This gene is located within the Beckwith-Wiedemann syndrome critical region-1 on chromosome 11p15. 5, and has been shown to be imprinted, with exclusive expression from the paternal allele.
Specifications
| TRPM5 | |
| Polyclonal | |
| Unconjugated | |
| trpm5 | |
| 9430099A16Rik; Long transient receptor potential channel 5; long transient receptor potential-related channel 5; LTRPC5; LTrpC-5; MLSN1 and TRP-related; MLSN1- and TRP-related gene 1 protein; MTR1; si:dkey-56p7.1; transient receptor potential cation channel subfamily M member 5; transient receptor potential cation channel, subfamily M, member 5; transient receptor potential melastatin 5; TRPM5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29850, 56843 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.84 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9JJH7, Q9NZQ8 | |
| trpm5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1026-1165 of human TRPM5 (NP_0553701). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.