Learn More
Invitrogen™ TRPM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534283
Description
Percent identity with other species by BLAST analysis: Human (100%); Chimpanzee, Gorilla, Orangutan, Monkey, Mouse (94%); Gibbon, Galago, Rat, Dog (88%); Marmoset, Elephant, Panda, Bovine, Pig (82%). Use in IHC with formalin-fixed paraffin-embedded tissues requires proteinase K digestion for antigen retrieval.
Calcium-activated non selective (CAN) cation channel that mediates membrane depolarization. While it is activated by increase in intracellular Ca2+, it is impermeable to it. Mediates transport of monovalent cations (Na+, K+, Cs+, Li+), leading to depolarize the membrane. It thereby plays a central role in cadiomyocytes, neurons from entorhinal cortex, dorsal root and vomeronasal neurons, endocrine pancreas cells, kidney epithelial cells, cochlea hair cells etc. Participates in T-cell activation by modulating Ca2+ oscillations after T lymphocyte activation, which is required for NFAT-dependent IL2 production. Involved in myogenic constriction of cerebral arteries. Controls insulin secretion in pancreatic beta-cells. May also be involved in pacemaking or could cause irregular electrical activity under conditions of Ca2+ overload. Affects T-helper 1 (Th1) and T-helper 2 (Th2) cell motility and cytokine production through differential regulation of calcium signaling and NFATC1 localization. Enhances cell proliferation through up-regulation of the beta-catenin signaling pathway.
Specifications
| TRPM4 | |
| Polyclonal | |
| Unconjugated | |
| TRPM4 | |
| 1110030C19Rik; AW047689; Calcium-activated non-selective cation channel 1; H LTrpC-4; hTRPM4; Long transient receptor potential channel 4; LOW QUALITY PROTEIN: transient receptor potential cation channel subfamily M member 4; LTrpC4; LTrpC-4; melastatin like 2 protein; melastatin-4; Melastatin-like 2; MLS2s; PFHB1B; transient receptor potential cation channel subfamily M member 4; transient receptor potential cation channel, subfamily M, member 4; transient receptor potential ion channel melastatin subgroup member 4; Trpm4; TRPM4B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8TD43 | |
| TRPM4 | |
| Synthetic 17 amino acid peptide from N-Terminus of human TRPM4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.