missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM4 Antibody (OTI10H5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-48036
This item is not returnable.
View return policy
Description
TRPM4 Monoclonal specifically detects TRPM4 in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRPM4 | |
| Monoclonal | |
| 0.77 mg/mL | |
| Western Blot 1:5000-10000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 | |
| TRPM4 | |
| Full length human recombinant protein of human TRPM4 (NP_060106) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 54795 | |
| Human, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI10H5 | |
| Unconjugated | |
| Calcium-activated non-selective cation channel 1, FLJ20041, hTRPM4, Long transient receptor potential channel 4, LTrpC-4, LTRPC4, melastatin-4, PFHB1B, transient receptor potential cation channel subfamily M member 4, transient receptor potential cation channel, subfamily M, member 4, TRPM4B | |
| Mouse | |
| 134.3 kDa | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for Homo sapiens transient receptor potential cation channel, subfamily M, member 4 (TRPM4). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction