Learn More
TRPM4 Antibody (CL11214) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07990C
Product Code. 30291187
  • £558.00 / 0.10mL
Description
TRPM4 Monoclonal antibody specifically detects TRPM4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPM4
Monoclonal
DyLight 650
Calcium-activated non-selective cation channel 1, FLJ20041, hTRPM4, Long transient receptor potential channel 4, LTrpC-4, LTRPC4, melastatin-4, PFHB1B, transient receptor potential cation channel subfamily M member 4, transient receptor potential cation channel, subfamily M, member 4, TRPM4B
This antibody was developed using a recombinant protein derived from Q8TD43, with the exact immunogen sequence remaining proprietary.
0.1 mL
Endocrinology, Neuroscience, Signal Transduction
54795
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL11214
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified