Learn More
Invitrogen™ TRPM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139988
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VDIRLAQLED LIGRMATALE RLTGLERAES NKIRSRTSSD CTDAAYIVRQ SSFNSQEGNT FKLQ Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
The product of this gene belongs to the family of transient receptor potential (TRP) channels. TRP channels are cation-selective channels important for cellular calcium signaling and homeostasis. The protein encoded by this gene mediates calcium entry, and this entry is potentiated by calcium store depletion. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| TRPM3 | |
| Polyclonal | |
| Unconjugated | |
| TRPM3 | |
| 6330504P12Rik; 9330180E14; AU018608; B930001P07Rik; GON-2; KI; KIAA1616; Long transient receptor potential channel 3; LTRPC3; LTrpC-3; melastatin 2; melastatin-2; MLSN2; si:dkey-201c13.4; transient receptor potential cation channel subfamily M member 3; transient receptor potential cation channel, subfamily M, member 3; transient receptor potential melastatin 3; transient receptor potential melastatin 3 beta 1; transient receptor potential melastatin 3 beta 10; transient receptor potential melastatin 3 beta 11; transient receptor potential melastatin 3 beta 12; transient receptor potential melastatin 3 beta 13; transient receptor potential melastatin 3 beta 14; transient receptor potential melastatin 3 beta 15; transient receptor potential melastatin 3 beta 16; transient receptor potential melastatin 3 beta 17; transient receptor potential melastatin 3 beta 2; transient receptor potential melastatin 3 beta 3; transient receptor potential melastatin 3 beta 4; transient receptor potential melastatin 3 beta 5; transient receptor potential melastatin 3 beta 6; transient receptor potential melastatin 3 beta 7; transient receptor potential melastatin 3 beta 8; transient receptor potential melastatin 3 beta 9; trpm3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80036 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HCF6 | |
| TRPM3 | |
| Recombinant protein corresponding to Human TRPM3. Recombinant protein control fragment (Product #RP-109408). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.