missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111058
This item is not returnable.
View return policy
Description
Immunogen sequence: RTRLEKFISE QTKERGGVAI KIPIVCVVLE GGPGTLHTID NATTNGTPCV VVEGSGRVAD VIAQVANLPV SDITISLIQQ KLSVFFQEMF ETFTESRIVE WTKKIQDIVR RRQLLTVFRE GKDGQQDV Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 94%.
TRPM2 belongs to the ion transport protein family which includes Sodium, Potassium, Calcium ion channels. This family is characterized by proteins possessing 6 transmembrane helices in which the last two helices flank a loop which determines ion selectivity. TRPM2 is activated by oxidative stress and confers susceptibility to cell death. Further, TRPM2 is a calcium-permeable cation channel that is regulated by free intracellular ADP-ribose. Several alternatively spliced transcript variants of the TRPM2 gene have been described, but their full-length nature is not known.
Specifications
| TRPM2 | |
| Polyclonal | |
| Unconjugated | |
| TRPM2 | |
| 9830168K16Rik; C79133; EC 3.6.1.13; EREG 1; EREG1; Estrogen-responsive element-associated gene 1 protein; KNP 3; KNP3; Long transient receptor potential channel 2; LTRPC 2; LTRPC2; LTrpC-2; MGC133383; NUDT9H; NUDT9L1; null; transient receptor potential cation channel subfamily M member 2; transient receptor potential cation channel, subfamily M, member 2; transient receptor potential channel 7; Transient receptor potential melastatin 2; transient receptor potential melastatin family 2; transient receptor protein 7; Trp7; TRPC 7; TRPC7; TRPM 2; Trpm2; Trpm2-predicted; Trrp7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7226 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.50 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94759 | |
| TRPM2 | |
| Recombinant Protein corresonding to Human TRPM2. Recombinant protein control fragment (Product #RP-103848). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction