missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPM2 Polyclonal Antibody, DyLight™ 550

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522880
This item is not returnable.
View return policy
Description
TRPM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)
TRPM2 belongs to the ion transport protein family which includes Sodium, Potassium, Calcium ion channels. This family is characterized by proteins possessing 6 transmembrane helices in which the last two helices flank a loop which determines ion selectivity. TRPM2 is activated by oxidative stress and confers susceptibility to cell death. Further, TRPM2 is a calcium-permeable cation channel that is regulated by free intracellular ADP-ribose. Several alternatively spliced transcript variants of the TRPM2 gene have been described, but their full-length nature is not known.
Specifications
| TRPM2 | |
| Polyclonal | |
| DyLight 550 | |
| TRPM2 | |
| 9830168K16Rik; C79133; EC 3.6.1.13; EREG 1; EREG1; Estrogen-responsive element-associated gene 1 protein; KNP 3; KNP3; Long transient receptor potential channel 2; LTRPC 2; LTRPC2; LTrpC-2; MGC133383; NUDT9H; NUDT9L1; null; transient receptor potential cation channel subfamily M member 2; transient receptor potential cation channel, subfamily M, member 2; transient receptor potential channel 7; Transient receptor potential melastatin 2; transient receptor potential melastatin family 2; transient receptor protein 7; Trp7; TRPC 7; TRPC7; TRPM 2; Trpm2; Trpm2-predicted; Trrp7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28240, 294329 | |
| 4°C, store in dark | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.83 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| E9PTA2, Q91YD4 | |
| TRPM2 | |
| Synthetic peptide made to a C-terminal portion of the rat TRPM2 protein (within residues 1430-1507). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction