missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPC7 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142465
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to cross-react with isoform 1, 2 and 3 (NP_065122.1; NP_001161049.1; NP_001161048.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.
Specifications
| TRPC7 | |
| Polyclonal | |
| Unconjugated | |
| TRPC7 | |
| capacitative calcium channel TRPC7; hTRP7; likley ortholog of mouse transient receptor potential cation channel, subfamily C, member 7; mTRP7; putative capacitative calcium channel; short transient receptor potential channel 7; transient receptor potential cation channel subfamily C member 7; transient receptor potential cation channel, subfamily C, member 7; transient receptor protein 7; TRP7; TRP-7; TRP7 protein; TRPC7; Trrp8 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 26946, 427641 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9WVC5 | |
| TRPC7 | |
| Peptide with sequence KLCKSKAKSCEND. | |
| 100 μg | |
| Primary | |
| Mouse, Chicken | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction