Learn More
TRPC6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-18236-20UL
Description
The TRPC6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRPC6. This antibody recognizes human, mouse antigen. The TRPC6 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| TRPC6 | |
| 1000 μg/mL | |
| FLJ11098, FLJ14863, FSGS2, Transient receptor protein 6, TRP 6, TRP6, TRPC6 | |
| 20 μL | |
| 7225 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |