missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TRPC5 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16864445
|
Proteintech
25890-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16854445
|
Proteintech
25890-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPC5 belongs to the transient receptor family. TRPC5 is one of the seven mammalian TRPC (transient receptor potential channel) proteins. The TRPC5 protein is a multi-pass membrane protein and is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC5 is active alone or as a heteromultimeric assembly with TRPC1, TRPC3, and TRPC4. TRPC5 also interacts with multiple proteins including calmodulin, CABP1, enkurin, Na(+)-H+ exchange regulatory factor (NHERF ), interferon-induced GTP-binding protein (MX1), ring finger protein 24 (RNF24), and SEC14 domain and spectrin repeat-containing protein 1 (SESTD1). TRPC5 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC5 is probably operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases, or G-protein coupled receptors. TRPC5 has also been shown to be calcium-selective, and may also be activated by intracellular calcium store depletion. Diseases associated with TRPC5 include Hypertrophic Pyloric Stenosis and Pyloric Stenosis.Specifications
| TRPC5 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q9QX29, Q9UL62 | |
| 22067, 7224 | |
| TRPC5 Fusion Protein Ag23171 | |
| Primary | |
| -20°C | |
| TRPC5 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| hTRP 5, hTRP5, Transient receptor protein 5, TRP 5, TRP5, TRPC5 | |
| TRPC5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title