missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPC4AP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111595
This item is not returnable.
View return policy
Description
Immunogen sequence: LIWRKHSASA LVLHGHNQNC DCSPDITLKI QFLRLLQSFS DHHENKYLLL NNQELNELSA ISLKANIPEV EAVLNTDRSL VCDGK Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control. The DCX(TRUSS) complex specifically mediates the polyubiquitination and subsequent degradation of MYC. Also participates in the activation of NFKB1 in response to ligation of TNFRSF1A, possibly by linking TNFRSF1A to the IKK signalosome. Involved in JNK activation via its interaction with TRAF2. Also involved in elevation of endoplasmic reticulum Ca(2+) storage reduction in response to CHRM1.
Specifications
| TRPC4AP | |
| Polyclonal | |
| Unconjugated | |
| TRPC4AP | |
| 4833429F06Rik; C20orf188; D2Ertd113e; hypothetical protein; mFLJ00177; PPP1R158; protein phosphatase 1, regulatory subunit 158; Protein TAP1; Protein TRUSS; Rabex-5/Rin2-interacting protein; short transient receptor potential channel 4 associated protein; short transient receptor potential channel 4-associated protein; TAP1 protein; TNF-receptor; TNF-receptor ubiquitous scaffolding/signaling protein; TP4AP; transient receptor potential cation channel subfamily C member 4 associated protein; transient receptor potential cation channel, subfamily C, member 4 associated protein; transient receptor protein 4, associated protein; TRP4-associated protein; Trp4-associated protein TAP1; Trpc4ap; trpc4-associated protein; Trrp4ap; TRUSS; tumor necrosis factor receptor-associated ubiquitous scaffolding and signaling protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26133 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TEL6 | |
| TRPC4AP | |
| Recombinant Protein corresonding to Human TRPC4AP. Recombinant protein control fragment (Product #RP-107400). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction