missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | TRPC3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TRPC3 Polyclonal specifically detects TRPC3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TRPC3 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CAA74083 | |
| 7222 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human TRPC3. The immunogen is located within amino acids 120 - 170 of TRPC3. Amino Acid Squence: FYAYDEDGTRFSPD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 100 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| hTrp3, hTrp-3, short transient receptor potential channel 3, transient receptor potential cation channel, subfamily C, member 3, transient receptor potential channel 3, Transient receptor protein 3, TRP3, TRP-3, TrpC3 | |
| TRPC3 | |
| IgG | |
| Affinity Purified | |
| This antibody may cross-react with TRPC6 and TRPC7. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title