missing translation for 'onlineSavingsMsg'
Learn More

TRPC3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18425273 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425273 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425273 Supplier Novus Biologicals Supplier No. NBP177262AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRPC3 Polyclonal antibody specifically detects TRPC3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPC3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias hTrp3, hTrp-3, short transient receptor potential channel 3, transient receptor potential cation channel, subfamily C, member 3, transient receptor potential channel 3, Transient receptor protein 3, TRP3, TRP-3, TrpC3
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human TRPC3. The immunogen is located within amino acids 760 - 810 of TRPC3. Amino Acid Squence: VDKENDEVNEGELK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7222
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.