missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£165.00 - £408.00
Specifications
| Concentration | LYOPH |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Antisera |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284344
|
Novus Biologicals
NB100-98844-0.025ML |
0.025 mL |
£165.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18727783
|
Novus Biologicals
NB100-98844 |
£408.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TRPC1 Polyclonal specifically detects TRPC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| P48995 | |
| 7220 | |
| IgG | |
| Unpurified | |
| TRPC1. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| HTRP-1, MGC133334, MGC133335, transient receptor potential canonical 1, transient receptor potential cation channel, subfamily C, member 1, transient receptor potential channel 1, Transient receptor protein 1, TRP-1, TRP1short transient receptor potential channel 1, TrpC1 | |
| TRPC1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title