missing translation for 'onlineSavingsMsg'
Learn More

TRPA1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 41 publication

£248.00 - £493.00

Specifications

Antigen TRPA1
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 28990934), ELISA reported in scientific literature (PMID 29803505), Immunohistochemistry 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:100-1:250, Immunohistochemistry-Frozen reported in scientific literature
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18776333
View Documents
Novus Biologicals
NB110-40763SS
0.025 mL
£248.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18728053
View Documents
Novus Biologicals
NB110-40763
0.1 mL
£493.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRPA1 Polyclonal specifically detects TRPA1 in Human, Mouse, Rat, Guinea Pig, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

TRPA1
Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 28990934), ELISA reported in scientific literature (PMID 29803505), Immunohistochemistry 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:100-1:250, Immunohistochemistry-Frozen reported in scientific literature
Polyclonal
Rabbit
Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems
PBS with 0.05% Sodium Azide
ANKTM1ankyrin-like with transmembrane domains 1, Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, transient receptor potential cation channel subfamily A member 1, transient receptor potential cation channel, subfamily A, member 1
TRPA1
IgG
Affinity Purified
Additional modified form of TRPA1 can also be detected.
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Guinea Pig
O75762
8989
A synthetic peptide made to a region within the N-terminus (residues 1-100) of the human TRPA1 protein. [Swiss-Prot# O75762]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.