missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRP7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99522-100ul
This item is not returnable.
View return policy
Description
TRP7 Polyclonal antibody specifically detects TRP7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TRP7 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| hTRP7, KNP3, likley ortholog of mouse transient receptor potential cation channel, subfamilyC, member 7, putative capacitative calcium channel, short transient receptor potential channel 7, transient receptor potential cation channel, subfamily C, member 7, transient receptor potential-related channel 7, a novel putative Ca2+ channelprotein10TRPM2, Transient receptor protein 7, TRP7, TRP-7, TrpC7 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TRP7. | |
| 100 μg | |
| Neuroscience, Signal Transduction | |
| 57113 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction