missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRP7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94388-0.1ml
This item is not returnable.
View return policy
Description
TRP7 Polyclonal antibody specifically detects TRP7 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TRP7 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| hTRP7, KNP3, likley ortholog of mouse transient receptor potential cation channel, subfamilyC, member 7, putative capacitative calcium channel, short transient receptor potential channel 7, transient receptor potential cation channel, subfamily C, member 7, transient receptor potential-related channel 7, a novel putative Ca2+ channelprotein10TRPM2, Transient receptor protein 7, TRP7, TRP-7, TrpC7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 720-780 of human TRPC7 (NP_065122.1). YLIMRIKMCLIKLCKSKAKSCENDLEMGMLNSKFKKTRYQAGMRNSENLTANNTLSKPTRY | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 57113 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction