missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£219.00 - £459.00
Specifications
| Antigen | TRP2 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18421191
|
Novus Biologicals
NBP1-86892-25ul |
25 μL |
£219.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221498
|
Novus Biologicals
NBP1-86892 |
0.1 mL |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRP2 Polyclonal antibody specifically detects TRP2 in Human, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
| TRP2 | |
| Unconjugated | |
| RUO | |
| Human, Canine, Equine | |
| dopachrome delta-isomerase, dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2), DT, EC 5.3.3.12, L-dopachrome tautomerase, TRP2, TRP-2, Tyrosinase-related protein 2, TYRP2, TYRP2L-dopachrome Delta-isomerase | |
| DCT | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1638 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NECDVCTDQLFGAARPDDPTLISRNSRFSSWETVCDSLDDYNHLVTLCNGTYEGLLRRNQMGRNSMKLPTLKDIRDCLSLQKFDNPPFFQNSTFSFRNALEGFDKADGTLDSQVMSLHNLVHSFLNGTNALPHSAANDPIFVVLHSFTDA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title