missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROY/TNFRSF19 Rabbit anti-Human, Mouse, Rat, Clone: 1R8Y5, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | TROY/TNFRSF19 |
|---|---|
| Clone | 1R8Y5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18356795
|
Novus Biologicals
NBP3-15699-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18353792
|
Novus Biologicals
NBP3-15699-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TROY/TNFRSF19 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 55504 | |
| IgG | |
| Affinity purified |
| 1R8Y5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-200 of human TROY/TNFRSF19 (Q9NS68). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title