missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROY/TNFRSF19 Antibody (SR1804), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | TROY/TNFRSF19 |
|---|---|
| Clone | SR1804 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615534
|
Novus Biologicals
NBP3-22336-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653194
|
Novus Biologicals
NBP3-22336-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| TROY/TNFRSF19 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| A synthesized peptide derived from human TROY/TNFRSF19 (Uniprot #: Q9NS68) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1804 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 55504 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title