missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TROY/TNFRSF19 Antibody Pair [HRP]

Product Code. p-7183038 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18270347

Brand: Novus Biologicals™ NBP27953615PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Mouse TROY/TNFRSF19.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.8 pg/mL
Assay Sensitivity 7.81-500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse TROY/TNFRSF19.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Gene ID (Entrez) 55504
Gene Symbol TNFRSF19
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 130 ng of recombinant mouse TROY/TNFRSF19.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-mouse TROY/TNFRSF19 monoclonal antibody.
  • Detection Antibody: 1 mg/mL rabbit anti-TROY/TNFRSF19 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis
    Storage Requirements Storage is content dependent.
    Target TROY/TNFRSF19
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.