missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROY/TNFRSF19 Antibody (933214), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB1548PE
This item is not returnable.
View return policy
Description
TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry.
Specifications
| TROY/TNFRSF19 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| TNFRSF19 | |
| Partial recombinant human TROY/TNFRSF protein (19 aa30-170) | |
| 100 μL | |
| Apoptosis | |
| 55504 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 933214 | |
| Flow Cytometry | |
| TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction