missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROY/TNFRSF19 Antibody (933202) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB15481JF585
This item is not returnable.
View return policy
Description
TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| TROY/TNFRSF19 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Mouse myeloma cell line NS0-derived recombinant human TROY/TNFRSF19, Glu30-Leu170, Accession # Q9NS68 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 933202 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis | |
| 55504 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction