missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROY/TNFRSF19 Antibody (208), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90451AF350
This item is not returnable.
View return policy
Description
TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for ELISA.
Specifications
| TROY/TNFRSF19 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human RELT Human DR6 Mouse FAS Mouse HVEM Mouse CD27 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 208 | |
| ELISA | |
| TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TROY/TNFRSF19 (NP_001157627.1; Met 1-Leu 170). | |
| 0.1 mL | |
| Apoptosis | |
| 55504 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction