missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TROY Monoclonal Antibody (6F5D3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538646
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
Tumor necrosis factor receptor superfamily member 19, commonly known as TROY, is a gene located on chromosome 1p36.22 that encodes a member of the tumor necrosis factor receptor (TNFR) superfamily. TROY is expressed in the nervous system, skin, and various tissues, where it plays a role in regulating cell apoptosis, inflammation, and development. As a receptor, TROY interacts with ligands and other signaling molecules to modulate pathways involved in cell survival, differentiation, and immune responses. In the context of the nervous system, TROY has been implicated in axonal outgrowth and regeneration, particularly influencing signaling pathways related to neural development and repair. Additionally, TROY has been studied in cancer research due to its potential role in tumor growth and progression, making it a candidate for therapeutic targeting. Its involvement in diverse biological processes highlights its significance in maintaining cellular homeostasis and its potential as a therapeutic target in neurobiology and oncology.
Specifications
| TROY | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9NS68 | |
| Tnfrsf19 | |
| Purified recombinant fragment of human TNFRSF19 (aa: extra 30-170) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 6F5D3 | |
| Unconjugated | |
| Tnfrsf19 | |
| AL023044; AW123854; RGD1564996; TAJ; TAJ-alpha; TNF receptor superfamily member 19; Tnfrsf19; Toxicity and JNK inducer; TRADE; TROY; tumor necrosis factor receptor superfamily member 19; tumor necrosis factor receptor superfamily, member 19; UNQ1888/PRO4333 | |
| Mouse | |
| Protein G | |
| RUO | |
| 55504 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction