missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troxerutin-d12 (>85%), TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Troxerutin-d12 |
| Molecular Formula | C33 D12 H30 O19 |
| Quantity | 5 mg |
| Synonym | 2-[3,4-Bis(2-hydroxethoxy)phenyl]-3-[[6-O-(6-O-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-7-(2-hydroxyethoxy)-4H-1-benzopyran-4-one-d12, 3',4',7-Tris(hydroxyethyl)rutin-d12, Posorutin-d12, Rufen-P4-d12, Ruven-d12 THR, Trioxyethylrutin-d12, Troxerutine-d12, Troxevazin-d12, Vastribil-d12, Veinamitol-d12, Veniten-d12, Venoruton P4-d12 |
| InChI Formula | InChI=1S/C33H42O19/c1-14-23(38)26(41)28(43)32(49-14)48-13-21-24(39)27(42)29(44)33(51-21)52-31-25(40)22-17(37)11-16(45-7-4-34)12-20(22)50-30(31)15-2-3-18(46-8-5-35)19(10-15)47-9-6-36/h2-3,10-12,14,21,23-24,26-29,32-39,41-44H,4-9,13H2,1H3/t14-,21?,23-,24+,26+,27-,28?,29+,32?,33-/m1/s1/i4D2,5D2,6D2,7D2,8D2,9D2 |
| SMILES | [2H]C([2H])(O)C([2H])([2H])Oc1cc(O)c2C(=O)C(=C(Oc2c1)c3ccc(OC([2H])([2H])C([2H])([2H])O)c(OC([2H])([2H])C([2H])([2H])O)c3)O[C@H]4OC(COC5O[C@H](C)[C@@H](O)[C@H](O)C5O)[C@H](O)[C@@H](O)[C@@H]4O |
| IUPAC Name | 2-[3,4-bis(1,1,2,2-tetradeuterio-2-hydroxyethoxy)phenyl]-5-hydroxy-7-(1,1,2,2-tetradeuterio-2-hydroxyethoxy)-3-[(2R,3S,4R,5R)-3,4,5-trihydroxy-6-[[(4S,5S,6R)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
| Molecular Weight (g/mol) | 754.749 |
| Formula Weight | 754.307 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction