missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROVE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12718-1-AP-20UL
This item is not returnable.
View return policy
Description
TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.Specifications
| TROVE2 | |
| Polyclonal | |
| Unconjugated | |
| TROVE2 | |
| 60 kDa ribonucleoprotein Ro, 60 kDa Ro protein, Ro 60 kDa autoantigen, RO60, RoRNP, Sjoegren syndrome antigen A2, SS A, SSA2, TROVE domain family member 2, TROVE domain family, member 2, TROVE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20822, 304833, 6738 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08848, P10155 | |
| TROVE2 | |
| TROVE2 Fusion Protein Ag3412 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction