missing translation for 'onlineSavingsMsg'
Learn More

Trovafloxacin, TRC

Product Code. 30134578
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134578 25 mg 25mg
30134418 2.5 mg 2.5mg
30011905 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134578 Supplier Toronto Research Chemicals Supplier No. TRCT89276025MG
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147059-72-1
Molecular Formula C20H15F3N4O3
Molecular Weight (g/mol) 416.35
Synonym 7-((1R,5S,6s)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl)-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-1,8-naphthyridine-3-carboxylic Acid, Turvel, 3-Azabicyclo[3.1.0]hexane, 1,8-naphthyridine-3-carboxylic acid derivative, 7-[(1alpha,5alpha,6alpha)-6-Amino-3-azabicyclo[3.1.0]hex-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic Acid
IUPAC Name 7-[(1R,5S)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES N[C@@H]1[C@H]2CN(C[C@@H]12)c3nc4N(C=C(C(=O)O)C(=O)c4cc3F)c5ccc(F)cc5F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H15F3N4O3/c21-8-1-2-15(13(22)3-8)27-7-12(20(29)30)17(28)9-4-14(23)19(25-18(9)27)26-5-10-11(6-26)16(10)24/h1-4,7,10-11,16H,5-6,24H2,(H,29,30)/t10-,11+,16+
Formula Weight 416.1096
Chemical Name or Material Trovafloxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.