missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Trouble-Shooter Cleaner, Aerosol
Description
- Cleans tough stains from difficult-to-reach areas
- Easy application with direction nozzle
- Clings to vertical surfaces without dripping
- Use with the 3M™ Doodlebug™ Cleaning System for best results
Specifications
Specifications
| Product Type | Floor Stripper |
| Form | Aerosol |
| Description | Cleaner |
| For Use With (Application) | Removing Heavy Soil, Wax, or Floor Finish Build-Ups From Hard Surfaces |
| For Use With (Equipment) | 3 M™ Doodlebug™ Cleaning System |
| Volume (Metric) | 600 mL |
Safety and Handling
Product Identifier
- 3M TroubleShooter Baseboard Stripper
- Danger
- Flammable and non-flammable aerosols category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 1 B
- H229-Pressurised container: May burst if heated.
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P251-Do not pierce or burn, even after use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
- MIXTURE LIST-Contain: 2-butoxyethanol, petroleum gases, isobutane, 2-aminoethanol,propane, alcohols C12-15 ethoxylated
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 9% of components with unknown hazards to the aquatic environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?