missing translation for 'onlineSavingsMsg'
Learn More

Trospium Chloride, TRC

Product Code. 30455151
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455151 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455151 Supplier Toronto Research Chemicals Supplier No. TRCT8928001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trospium chloride
CAS 10405-02-4
Molecular Formula C25 H30 N O3 . Cl
Quantity 1 g
Synonym Trospium chloride, (1R,3r,5S)-3-[(Hydroxydiphenylacetyl)oxy]spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] chloride, Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium]','3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-','chloride (1:1)','(1α,3β,5α)-, Spiro[1αH,5αH-nortropane-8,1'-pyrrolidinium]','3α-hydroxy-','chloride','benzilate (8CI), Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium]','3-[(hydroxydiphenylacetyl)oxy]-','chloride','(1α,3β,5α)- (9CI), Benzilic acid','ester with 3α-hydroxyspiro[1αH,5αH-nortropane-8,1'-pyrrolidinium] chloride (8CI), As XVII, Azoniaspiro compound XVII, Azoniaspiro(3α-benziloyloxynortropane-8,1'-pyrrolidine) chloride, Keptan, Relaspium, Sanctura, Sanctura XR, Spasmex, Spasmo 3, Spasmo-lyt, Trospium chloride, Trozyd
InChI Formula InChI=1S/C25H30NO3.ClH/c27-24(25(28,19-9-3-1-4-10-19)20-11-5-2-6-12-20)29-23-17-21-13-14-22(18-23)26(21)15-7-8-16-26;/h1-6,9-12,21-23,28H,7-8,13-18H2;1H/q+1;/p-1/t21-,22+,23+;
SMILES [Cl-].OC(C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)[N+]23CCCC3)(c4ccccc4)c5ccccc5
IUPAC Name [(1S,5R)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride
Molecular Weight (g/mol) 427.96
Formula Weight 427.1914
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.