missing translation for 'onlineSavingsMsg'
Learn More

Trospium chloride, MedChemExpress

Product Code. 16418326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418326 Supplier Medchem Express Supplier No. HYB0461
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trospium chloride is an orally active, specific and competitive antagonist of muscarinic cholinergic receptors (mAChRs), with antimuscarinic activity. Trospium chloride binds to muscarinic receptors M1, M2 and M3 with high affinity, but not nicotinic, cholinergic receptors.

Trospium chloride is an orally active, specific and competitive antagonist of muscarinic cholinergic receptors (mAChRs), with antimuscarinic activity. Trospium chloride binds to muscarinic receptors M1, M2 and M3 with high affinity, but not nicotinic, cholinergic receptors.

Specifications

Chemical Name or Material Trospium chloride
Physical Form Solid
CAS 10405-02-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H30ClNO3
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : ≥ 100 mg/mL (233.67 mM) ∣DMSO : 33.33 mg/mL (77.88 mM; Need ultrasonic)
SMILES [H][C@@]12CC[C@@](C[C@@H](OC(C(C3=CC=CC=C3)(O)C4=CC=CC=C4)=O)C2)([H])[N+]15CCCC5.[Cl-]
Molecular Weight (g/mol) 427.96
Formula Weight 427.96
Percent Purity 99.65%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.