missing translation for 'onlineSavingsMsg'
Learn More

Trospium Chloride, LoGiCal

Product Code. 30463491
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463491 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463491 Supplier LoGiCal Supplier No. LGCFOR1266.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trospium chloride
CAS 10405-02-4
Analyte or Component Names Trospium chloride
Shipping Condition Room Temperature
Molecular Formula C25 H30 N O3 . Cl
Certifications/Compliance ISO 9001
Synonym Trospium chloride, (1R,3r,5S)-3-[(Hydroxydiphenylacetyl)oxy]spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] chloride, As XVII, Azoniaspiro compound XVII, Keptan, Relaspium, Sanctura, Sanctura XR, Spasmex, Spasmo 3, Spasmo-lyt, Trozyd, Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium], 3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-, chloride (1:1), (1alpha,3beta,5alpha)-, Spiro[1alphaH,5alphaH-nortropane-8,1'-pyrrolidinium], 3alpha-hydroxy-, chloride, benzilate (8CI), Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium], 3-[(hydroxydiphenylacetyl)oxy]-, chloride, (1alpha,3beta,5alpha)- (9CI), Benzilic acid, ester with 3alpha-hydroxyspiro[1alphaH,5alphaH-nortropane-8,1'-pyrrolidinium] chloride (8CI), Azoniaspiro(3alpha-benziloyloxynortropane-8,1'-pyrrolidine) chloride
InChI Formula InChI=1S/C25H30NO3.ClH/c27-24(25(28,19-9-3-1-4-10-19)20-11-5-2-6-12-20)29-23-17-21-13-14-22(18-23)26(21)15-7-8-16-26;/h1-6,9-12,21-23,28H,7-8,13-18H2;1H/q+1;/p-1/t21-,22+,23+;
Solution Type Neat
SMILES [Cl-].OC(C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)[N+]23CCCC3)(c4ccccc4)c5ccccc5
IUPAC Name [(1S,5R)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 427.96
Quantity 10 mg
Formula Weight 427.1914 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.