missing translation for 'onlineSavingsMsg'
Learn More

Tropylium tetrafluoroborate, 97%

Product Code. 11447946
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447946 100 g 100g
11427946 5 g 5g
11437946 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447946 Supplier Thermo Scientific Alfa Aesar Supplier No. A14045.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27081-10-3 | C7H7BF4 | 177.94 g/mol

Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.

Solubility
Soluble in hot methanol.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, strong bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27081-10-3
Molecular Formula C7H7BF4
Molecular Weight (g/mol) 177.94
MDL Number MFCD00013402
InChI Key SQVQHTIKOZVGLR-UHFFFAOYSA-N
Synonym tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t
PubChem CID 152926
IUPAC Name cyclohepta-1,3,5-triene;tetrafluoroborate
SMILES F[B-](F)(F)F.[CH+]1C=CC=CC=C1

Specifications

Melting Point ∼240°C (decomposition)
Quantity 100 g
UN Number UN1759
Beilstein 3574512
Sensitivity Moisture sensitive
Solubility Information Soluble in hot methanol.
Formula Weight 177.94
Percent Purity 97%
Chemical Name or Material Tropylium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.