missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin C (cardiac) Rabbit anti-Mouse, Rat, Clone: 8Z4V9, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | Troponin C (cardiac) |
|---|---|
| Clone | 8Z4V9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313412
|
Novus Biologicals
NBP3-16280-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18335684
|
Novus Biologicals
NBP3-16280-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Troponin C (cardiac) Monoclonal antibody specifically detects Troponin C (cardiac) in Mouse, Rat samples. It is validated for Western BlotSpecifications
| Troponin C (cardiac) | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 7134 | |
| IgG | |
| Affinity purified |
| 8Z4V9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Troponin C (cardiac) (P63316). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title