Learn More
Troponin C (cardiac) Antibody (OTI5H3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74638AF405
Product Code. 18127235
  • £595.00 / 0.10mL
Description
Troponin C (cardiac) Monoclonal specifically detects Troponin C (cardiac) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Troponin C (cardiac)
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
TNNC1
Full length recombinant protein of human Troponin C (cardiac) (NP_003271) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Immunohistochemistry
OTI5H3
Immunohistochemistry
cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow
Mouse
Affinity Purified
Cellular Markers, Cytoskeleton Markers
7134.0
Store at 4°C in the dark.