missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tropomyosin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48864-25ul
This item is not returnable.
View return policy
Description
tropomyosin-3 Polyclonal antibody specifically detects tropomyosin-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| tropomyosin-3 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| cytoskeletal tropomyosin TM30, FLJ41118, gamma-tropomyosin, heat-stable cytoskeletal protein 30 kDa, hscp30, hTM5, MGC14582, MGC3261, MGC72094, NEM1, OK/SW-cl.5, TM3, TM30, TM30nm, TM-5, TPMsk3, TRK, tropomyosin 3, tropomyosin alpha-3 chain, tropomyosin gamma, tropomyosin-3, Tropomyosin-5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EMQLKEAKHIAEEADRKYEEVARKLVILE | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7170 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction