missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropomodulin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79572
This item is not returnable.
View return policy
Description
Tropomodulin 2 Polyclonal specifically detects Tropomodulin 2 in Human samples. It is validated for Western Blot.
Specifications
| Tropomodulin 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MGC39481, Neuronal tropomodulin, N-Tmod, NTMODtropomodulin-2, tropomodulin 2 (neuronal) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Chicken: 92%; Rabbit: 92%; Zebrafish: 83%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_055363 | |
| TMOD2 | |
| Synthetic peptide directed towards the N terminal of human TMOD2. Peptide sequence MSSSPLSKKRRVSGPDPKPGSNCSPAQSVLSEVPSVPTNGMAKNGSEADI. | |
| 100 μL | |
| Neuroscience | |
| 29767 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction