missing translation for 'onlineSavingsMsg'
Learn More

Tropolone, 98%

Product Code. 10276913 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10276913 25 g Glass bottle 25g
10615962 1 g Glass bottle 1g
10533881 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10276913 Supplier Thermo Scientific Acros Supplier No. 140580250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 533-75-5 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-75-5
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00004158
InChI Key MDYOLVRUBBJPFM-UHFFFAOYSA-N
Synonym tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins
PubChem CID 10789
ChEBI CHEBI:79966
IUPAC Name 2-hydroxycyclohepta-2,4,6-trien-1-one
SMILES C1=CC=C(C(=O)C=C1)O

Specifications

Melting Point 50.0°C to 54.0°C
Color Beige-Cream to White
Boiling Point 115.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9963
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethyl ether, acetone, organic solvents
Formula Weight 122.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tropolone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.