missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tropisetron hydrochloride

Description
Potent, orally active 5-HT3 receptor antagonist. Antiemetic. Also partial agonist of α7 nAChR. Activates α7Q79G-GlyR chimeric ion channels (EC50 = 38 nM)
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 320.82 |
| Purity | >99% |
| Molecular Weight (g/mol) | 320.817 |
| Chemical Name or Material | Tropisetron hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?