missing translation for 'onlineSavingsMsg'
Learn More

Tropisetron Hydrochloride, MedChemExpress

Product Code. 16474207
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474207 50 mg 50mg
16438066 100 mg 100mg
16418328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474207 Supplier Medchem Express Supplier No. HYB0020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tropisetron Hydrochloride (SDZ-ICS-930) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.

Tropisetron Hydrochloride (SDZ-ICS-930) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.

Tekniske data

Chemical Name or Material Tropisetron Hydrochloride
Physical Form Solid
CAS 105826-92-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H21ClN2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym SDZ-ICS-930
Solubility Information H2O : ≥ 50 mg/mL (155.86 mM) ∣DMSO : 33.33 mg/mL (103.89 mM; Need ultrasonic)
SMILES O=C(C1=CNC2=C1C=CC=C2)O[C@H]3C[C@H]4CC[C@H](N4C)C3.[H]Cl
Molecular Weight (g/mol) 320.81
Formula Weight 320.81
Percent Purity 99.95%
Grade Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.