missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropisetron-d5, TRC
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | Tropisetron-d5 |
| Molecular Formula | C17H15D5N2O2 |
| Quantity | 25 mg |
| Synonym | 1H-Indole-3-carboxylic Acid (3-endo)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl Ester-d5, Novaban-d5, SDZ-ICS 930-d5 |
| InChI Formula | InChI=1S/C17H20N2O2/c1-19-11-6-7-12(19)9-13(8-11)21-17(20)15-10-18-16-5-3-2-4-14(15)16/h2-5,10-13,18H,6-9H2,1H3/t11-,12+,13?/i2D,3D,4D,5D,10D |
| SMILES | CN1[C@H]2CC[C@@H]1C[C@H](OC(C3=C([2H])NC4=C3C([2H])=C([2H])C([2H])=C4[2H])=O)C2 |
| Molecular Weight (g/mol) | 289.38 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction