missing translation for 'onlineSavingsMsg'
Learn More

Tropine, 98%

Product Code. 11387665
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387665 10 g 10g
11397665 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387665 Supplier Thermo Scientific Alfa Aesar Supplier No. L02332.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-29-6 | C8H15NO | 141.214 g/mol

Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

Solubility
Soluble in water. (100 g/L) at 20°C.

Notes
Hygroscopic. Store at 4°C. Incomaptible with Strong acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-29-6
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00005551
InChI Key CYHOMWAPJJPNMW-RNFRBKRXSA-N
Synonym tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
PubChem CID 6101956
IUPAC Name (1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
SMILES CN1C2CCC1CC(C2)O

Specifications

Melting Point 61°C to 65°C (dried)
Density 1.04
pH 12
Boiling Point 232°C to 234°C
Odor Amine-like
Quantity 10 g
Beilstein 80188
Sensitivity Hygroscopic
Merck Index 14,9781
Solubility Information Soluble in water. (100g/L) at 20°C.
Formula Weight 141.22
Percent Purity 98%
Chemical Name or Material Tropine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.