missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Tropine |
| CAS | 120-29-6 |
| Analyte or Component Names | Tropine |
| Shipping Condition | +20°C |
| Molecular Formula | C8 H15 N O |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Tropisetron Hydrochloride Imp. A (EP), Tropisetron Imp. A (EP), (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-ol, Tropine, Tropisetron Hydrochloride Impurity A, Tropisetron Impurity A |
| InChI Formula | InChI=1S/C8H15NO/c1-9-6-2-3-7(9)5-8(10)4-6/h6-8,10H,2-5H2,1H3/t6-,7+,8+ |
| SMILES | CN1[C@@H]2CC[C@H]1C[C@@H](O)C2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction