missing translation for 'onlineSavingsMsg'
Learn More

Tropicamide, Tocris Bioscience™

Product Code. 30001145
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30001145

Brand: Tocris Bioscience 0909/100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective M4 antagonist

Tropicamide is an M4 selective muscarinic receptor antagonist.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1508-75-4
Molecular Formula C17H20N2O2
Quantity 100 mg
Synonym tropicamide, mydriacyl, mydriaticum, mydriafair, tropicacyl, minims tropicamide, bistropamide, epitromina, tropicamidum, mydrum
InChI Key BGDKAVGWHJFAGW-UHFFFAOYSA-N
SMILES CCN(CC1=CC=NC=C1)C(=O)C(CO)C2=CC=CC=C2
IUPAC Name N-ethyl-3-hydroxy-2-phenyl-N-(pyridin-4-ylmethyl)propanamide
Molecular Weight (g/mol) 284.359
PubChem CID 5593
Formula Weight 284.36
Percent Purity >98%
Chemical Name or Material Tropicamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.