Learn More
Invitrogen™ Trop2 (EGP-1) Monoclonal Antibody (MR54), PE, eBioscience™

Mouse Monoclonal Antibody
£244.00
Specifications
| Antigen | Trop2 (EGP-1) |
|---|---|
| Clone | MR54 |
| Concentration | 5 μL/Test |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: This MR54 monoclonal antibody reacts with human Trop-2, which is also known as Tumor-Associated Calcium Signal Transducer 2 (TACSTD2) and EGP-1. This type I transmembrane glycoprotein is expressed on trophoblast (syncytiotrophoblast and cytotrophoblast), epithelial, and most human carcinoma cells. Trop-2 plays a role in calcium-mediated signaling. Trop-2 has also been reported as an oncogene, with expression being correlated with tumorigenesis, metastasis, and anchorage-independent growth. Applications Reported: This MR54 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR54 antibody has been pre-titrated and tested by flow cytometric analysis of MCF7 cell line. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
TROP2 is a carcinoma-associated antigen defined by the monoclonal antibody GA733. This antigen is a member of a family including at least two type I membrane proteins. It transduces an intracellular calcium signal and acts as a cell surface receptor. Mutations of its gene result in gelatinous drop-like corneal dystrophy, an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness.Specifications
| Trop2 (EGP-1) | |
| 5 μL/Test | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P09758 | |
| 4070 | |
| Primary | |
| Affinity chromatography |
| MR54 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human | |
| TACSTD2 | |
| 40kD glycoprotein, identified by monoclonal antibody GA733; C80403; cell surface glycoprotein TROP2; cell surface glycoprotein Trop-2; EGP1; EGP-1; epithelial glycoprotein-1; GA733; GA7331; GA733-1; gastrointestinal tumor-associated antigen GA7331; GP50; Ly97; lymphocyte antigen 97; M1S1; Membrane component chromosome 1 surface marker 1; membrane component, chromosome 1, surface marker 1; pancreatic carcinoma marker protein GA7331; pancreatic carcinoma marker protein GA733-1; parturition-related protein 1; Prp1; TACD2; TACSTD2; Trop2; truncated TACSTD2; tumor-associated calcium signal transducer 2; tunor-associated calcium signal transducer 2 | |
| TACSTD2 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.