missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Antibody (TACSTD2/2151), Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £392.00
Specifications
| Antigen | TROP-2 |
|---|---|
| Clone | TACSTD2/2151 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624538
|
Novus Biologicals
NBP2-75743-100ug |
100 μg |
£392.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618206
|
Novus Biologicals
NBP2-75743-20ug |
20 μg |
£232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TROP-2 Monoclonal antibody specifically detects TROP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| TROP-2 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| Recombinant fragment of human TROP-2 protein (around aa 31-274) (exact sequence is proprietary) (Uniprot: P09758 ) | |
| Primary | |
| Store at 4°C. |
| TACSTD2/2151 | |
| Western Blot 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P09758, P09758, P09758 | |
| 4070 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title