missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Antibody (MM0588-49D6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11982
This item is not returnable.
View return policy
Description
TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| TROP-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, CyTOF-ready | |
| P09758 | |
| TACSTD2 | |
| Human recombinant TROP-2 | |
| 0.1 mg | |
| Signal Transduction | |
| 4070 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| MM0588-49D6 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction