missing translation for 'onlineSavingsMsg'
Learn More

TROP-2 Antibody (30), Alexa Fluor™ 405 , Novus Biologicals™

Product Code. 18423298 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423298 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423298 Supplier Novus Biologicals Supplier No. NBP289494AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TROP-2 Monoclonal antibody specifically detects TROP-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TROP-2
Applications ELISA
Classification Monoclonal
Clone 30
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TROP-2 extracellular domain (NP_002344.2; Met 1-Thr 274).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4070
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.