missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Trontinemab Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555015
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Compatible with Direct ELISA.
Trontinemab is a monoclonal antibody that targets and neutralizes specific proteins involved in disease processes, thereby modulating the immune response.
Specifikationer
| Trontinemab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AD1TFRC | |
| Recombinant Human APP and CD71/TFRC. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 2.5 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction